The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025399.3 | NP_079675.1 | 3 - 4 | 240 | 68 |
GenBank mRNA | AK003991.1 | - | 3 - 4 | 247 | 68 |
AK013174.1 | - | 3 - 4 | 196 | 68 | |
AK184511.1 | - | 2 - 3 | 69 | ||
BC025444.1 | - | 3 - 4 | 242 | 68 |