The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133673.3 | NP_598434.2 | 1 - 2 | 222 | 73 |
GenBank mRNA | AJ297743.1 | - | 1 - 2 | 252 | 73 |
AK017290.1 | - | 1 - 2 | 241 | 73 | |
AK149754.1 | - | 1 - 2 | 243 | 73 | |
AK159843.1 | - | 1 - 2 | 221 | 73 | |
AK169692.1 | - | 1 - 2 | 245 | 73 | |
BC018456.1 | - | 1 - 2 | 221 | 73 |