Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gp6
Entrez Gene ID
243816
Gene Name
glycoprotein 6 (platelet)
Gene Aliases
9830166G18Rik, Gm469, Gpvi
Chromosome Location
Chr.7: 4362853 - 4397905 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163014.1 NP_001156486.1 1 - 2 97 61
GenBank mRNA AK079424.1 - 1 - 2 74 61
BC138170.1 - 1 - 2 81 61
BC145172.1 - 1 - 2 81 61

View Full Product Details