Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myh1
Entrez Gene ID
17879
Gene Name
myosin, heavy polypeptide 1, skeletal muscle, adult
Gene Aliases
A530084A17Rik, IId, IId/x, MHC-2X/D, MHC2X/D, MYHC-IIX, MdMs, MyHC-IId/x, MyHC-IIx/d, Myhs-f, Myhs-f2, Myhsf2
Chromosome Location
Chr.11: 67200114 - 67224575 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030679.1 NP_109604.1 28 - 29 4077 93
GenBank mRNA BC108329.1 - 28 - 29 4077 93

View Full Product Details