The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030679.1 | NP_109604.1 | 9 - 10 | 983 | 155 |
GenBank mRNA | AK041122.1 | - | 10 - 11 | 996 | 155 |
BC108329.1 | - | 9 - 10 | 983 | 155 |