The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010855.3 | NP_034985.2 | 34 - 35 | 5078 | 63 |
GenBank mRNA | AJ278733.1 | - | 4 - 5 | 722 | 63 |
BC141362.1 | - | 32 - 33 | 4995 | 63 |