Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myh2
Entrez Gene ID
17882
Gene Name
myosin, heavy polypeptide 2, skeletal muscle, adult
Gene Aliases
MHC2A, MyHC-IIa, Myh2a, Myhs-f, Myhs-f1, Myhsf1
Chromosome Location
Chr.11: 67171027 - 67197517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039545.2 NP_001034634.2 28 - 29 4005 80

View Full Product Details