The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039545.2 | NP_001034634.2 | 39 - 40 | 5799 | 103 |
GenBank mRNA | BC008538.1 | - | 37 - 38 | 5221 | 103 |