Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kdm6b
Entrez Gene ID
216850
Gene Name
KDM1 lysine (K)-specific demethylase 6B
Gene Aliases
1700064E03Rik, BC038313, Jmjd3, mKIAA0346
Chromosome Location
Chr.11: 69398508 - 69418148 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017426.1 NP_001017426.1 18 - 19 4683 71
GenBank mRNA AK129121.1 - 8 - 9 1475 71
AK136019.1 - 8 - 9 864 71
BC038313.1 - 3 - 4 321 71
BC075632.1 - 19 - 20 4859 71

View Full Product Details