Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kdm6b
Entrez Gene ID
216850
Gene Name
KDM1 lysine (K)-specific demethylase 6B
Gene Aliases
1700064E03Rik, BC038313, Jmjd3, mKIAA0346
Chromosome Location
Chr.11: 69398508 - 69418148 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017426.1 NP_001017426.1 6 - 7 864 63
GenBank mRNA BC075632.1 - 7 - 8 1040 63

View Full Product Details