The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110214.1 | NP_001103684.1 | 6 - 7 | 554 | 94 |
NM_146114.3 | NP_666226.2 | 6 - 7 | 554 | 94 | |
NM_175683.4 | NP_783614.1 | 6 - 7 | 554 | 94 | |
GenBank mRNA | AF387731.1 | - | 6 - 7 | 536 | 94 |
AK037126.1 | - | 6 - 7 | 521 | 94 | |
AK038788.1 | - | 6 - 7 | 484 | 94 | |
AK052369.1 | - | 6 - 7 | 510 | 94 | |
AK052490.1 | - | 6 - 7 | 493 | 94 | |
AK166324.1 | - | 6 - 7 | 94 | ||
BC108935.2 | - | 6 - 7 | 507 | 94 |