Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Evl
Entrez Gene ID
14026
Gene Name
Ena-vasodilator stimulated phosphoprotein
Gene Aliases
AI528774
Chromosome Location
Chr.12: 108554720 - 108688516 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163395.1 NP_001156867.1 1 - 2 246 74
GenBank mRNA AK134563.1 - 1 - 2 246 74

View Full Product Details