The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007556.2 | NP_031582.1 | 4 - 5 | 1407 | 119 |
GenBank mRNA | AK041210.1 | - | - | 1765 | 119 |
AK135770.1 | - | 4 - 5 | 1040 | 119 | |
BC138593.1 | - | 4 - 5 | 1327 | 119 | |
BC138595.1 | - | 4 - 5 | 1327 | 119 | |
J04566.1 | - | 4 - 5 | 989 | 119 | |
X80992.1 | - | 4 - 5 | 1406 | 119 |