Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbx3
Entrez Gene ID
12417
Gene Name
chromobox 3
Gene Aliases
HP1g, M32
Chromosome Location
Chr.6: 51470616 - 51483704 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC110376.1 - 1 - 2 79 140

View Full Product Details