Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syk
Entrez Gene ID
20963
Gene Name
spleen tyrosine kinase
Gene Aliases
Sykb
Chromosome Location
Chr.13: 52582329 - 52648792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198977.1 NP_001185906.1 13 - 14 2025 76
NM_011518.2 NP_035648.2 13 - 14 2029 76
GenBank mRNA AK136363.1 - 13 - 14 2028 76
AK143564.1 - 13 - 14 2058 76
BC065121.1 - 13 - 14 1961 76
U25685.1 - 12 - 13 1813 76
U36776.1 - 13 - 14 2032 76
Z49877.1 - 13 - 14 2287 76

View Full Product Details