Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Entpd6
- Entrez Gene ID
-
12497
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 6
- Gene Aliases
-
2700026H11Rik, Cd39l2, NTPDase-6
- Chromosome Location
-
Chr.2: 150749038 - 150771675 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_172117.5 |
NP_742115.2 |
10 - 11 |
1150 |
67 |
View Full Product Details