Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abhd12
Entrez Gene ID
76192
Gene Name
abhydrolase domain containing 12
Gene Aliases
1500011G07Rik, 6330583M11Rik, AI431047, AW547313
Chromosome Location
Chr.2: 150832493 - 150904745 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024465.3 NP_077785.2 7 - 8 932 79
GenBank mRNA AK018261.1 - 7 - 8 866 79
AK160126.1 - 7 - 8 932 79
AK219355.1 - 1 - 2 87 79
BC002138.1 - 7 - 8 901 79
BC002263.1 - 3 - 4 235 79

View Full Product Details