The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146025.1 | NP_001139497.1 | 8 - 9 | 1372 | 114 |
NM_001146026.1 | NP_001139498.1 | 8 - 9 | 1297 | 114 | |
NM_001146027.1 | NP_001139499.1 | 8 - 9 | 1163 | 114 | |
NM_134064.2 | NP_598825.2 | 8 - 9 | 1305 | 114 | |
NR_027395.1 | 8 - 9 | 1225 | 114 | ||
NR_027396.1 | 8 - 9 | 1222 | 114 | ||
GenBank mRNA | AK147349.1 | - | 8 - 9 | 1298 | 114 |
AK160090.1 | - | 8 - 9 | 1156 | 114 | |
AK160115.1 | - | 8 - 9 | 1156 | 114 | |
AK161651.1 | - | 8 - 9 | 1285 | 116 | |
AK171943.1 | - | 8 - 9 | 1154 | 114 | |
AK172257.1 | - | 8 - 9 | 1155 | 114 | |
BC017630.1 | - | 8 - 9 | 1098 | 114 | |
BC035548.1 | - | 8 - 9 | 1120 | 114 |