The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033669.2 | NP_001028841.2 | 10 - 11 | 2019 | 71 |
NM_001291441.1 | NP_001278370.1 | 10 - 11 | 2019 | 71 | |
GenBank mRNA | AK132680.1 | - | 10 - 11 | 2020 | 71 |
BC141212.1 | - | 10 - 11 | 2010 | 71 |