The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001602.2 | NP_001001602.2 | 11 - 12 | 3178 | 61 |
NM_001114124.2 | NP_001107596.1 | 13 - 14 | 3302 | 61 | |
NM_001114125.1 | NP_001107597.1 | 13 - 14 | 3154 | 61 | |
NM_001290637.1 | NP_001277566.1 | 13 - 14 | 3209 | 61 | |
NM_001290639.1 | NP_001277568.1 | 11 - 12 | 2837 | 61 | |
NM_001290640.1 | NP_001277569.1 | 11 - 12 | 3142 | 61 | |
NM_001290641.1 | NP_001277570.1 | 13 - 14 | 3435 | 61 | |
GenBank mRNA | AK147464.1 | - | 13 - 14 | 3234 | 61 |
AK147593.1 | - | 13 - 14 | 3436 | 61 | |
AK155296.1 | - | 13 - 14 | 3210 | 61 | |
AY178784.1 | - | 3 - 4 | 1070 | 61 | |
AY305656.1 | - | 11 - 12 | 2895 | 61 | |
AY305657.1 | - | 11 - 12 | 3178 | 61 | |
AY305658.1 | - | 11 - 12 | 2837 | 61 | |
BC118530.1 | - | 11 - 12 | 2803 | 61 | |
DQ473307.1 | - | 13 - 14 | 3039 | 61 |