Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Npdc1
Entrez Gene ID
18146
Gene Name
neural proliferation, differentiation and control 1
Gene Aliases
AI314472, NPDC-1
Chromosome Location
Chr.2: 25403050 - 25409494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC023887.1 - 1 - 2 552 98

View Full Product Details