The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146087.1 | NP_001139559.1 | 1 - 2 | 171 | 64 |
GenBank mRNA | AK014576.2 | - | - | 171 | 64 |