Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc34a3
Entrez Gene ID
142681
Gene Name
solute carrier family 34 (sodium phosphate), member 3
Gene Aliases
AI649385, Npt2c, Npt2c-v1, NptIIc, naPi-2c
Chromosome Location
Chr.2: 25228897 - 25234271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080854.3 NP_543130.2 4 - 5 399 73
GenBank mRNA AB054999.1 - 4 - 5 379 73
BC125326.1 - 3 - 4 332 73
BC131997.1 - 3 - 4 332 73
CJ068489.1 - 4 - 5 393 73

View Full Product Details