Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Slc34a3
- Entrez Gene ID
-
142681
- Gene Name
- solute carrier family 34 (sodium phosphate), member 3
- Gene Aliases
-
AI649385, Npt2c, Npt2c-v1, NptIIc, naPi-2c
- Chromosome Location
-
Chr.2: 25228897 - 25234271 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CJ068489.1 |
- |
2 - 3 |
174 |
76 |
View Full Product Details