Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Barhl1
Entrez Gene ID
54422
Gene Name
BarH-like 1 (Drosophila)
Gene Aliases
Dres115, MBH2
Chromosome Location
Chr.2: 28907680 - 28918415 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF264026.1 - 1 - 2 148 94

View Full Product Details