Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rmi1
Entrez Gene ID
74386
Gene Name
RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae)
Gene Aliases
4932432N11Rik, C79893
Chromosome Location
Chr.13: 58402248 - 58411149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168248.1 NP_001161720.1 2 - 3 284 201
GenBank mRNA BC037694.1 - 2 - 3 180 198

View Full Product Details