The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284410.2 | NP_001271339.1 | 1 - 2 | 231 | 71 |
GenBank mRNA | BC058175.1 | - | 1 - 2 | 231 | 71 |