The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198872.1 | NP_001185801.1 | 8 - 9 | 716 | 68 |
NM_001198873.1 | NP_001185802.1 | 8 - 9 | 701 | 68 | |
NM_001198874.1 | NP_001185803.1 | 6 - 7 | 647 | 68 | |
NM_001198875.1 | NP_001185804.1 | 6 - 7 | 605 | 68 | |
NM_001198876.1 | NP_001185805.1 | 5 - 6 | 587 | 68 | |
NM_001198877.1 | NP_001185806.1 | 8 - 9 | 705 | 68 | |
NM_001198878.1 | NP_001185807.1 | 8 - 9 | 690 | 68 | |
NM_010064.4 | NP_034194.1 | 5 - 6 | 576 | 68 | |
GenBank mRNA | AF063231.1 | - | 5 - 6 | 515 | 68 |
AK088015.1 | - | 5 - 6 | 547 | 68 | |
AK141837.1 | - | 6 - 7 | 615 | 68 | |
AK150173.1 | - | 5 - 6 | 537 | 68 | |
AK161831.1 | - | 1 - 2 | 1237 | 68 | |
AK164322.1 | - | 6 - 7 | 571 | 68 | |
AK164437.1 | - | 5 - 6 | 511 | 68 | |
AK168730.1 | - | 5 - 6 | 551 | 68 | |
BC139374.1 | - | 5 - 6 | 471 | 68 | |
BC139375.1 | - | 5 - 6 | 471 | 68 | |
GU992208.1 | - | 8 - 9 | 607 | 68 | |
GU992209.1 | - | 8 - 9 | 592 | 68 | |
GU992210.1 | - | 6 - 7 | 496 | 68 | |
GU992211.1 | - | 8 - 9 | 645 | 68 | |
GU992212.1 | - | 8 - 9 | 630 | 68 |