The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289487.1 | NP_001276416.1 | 6 - 7 | 1698 | 73 |
NM_001289488.1 | NP_001276417.1 | 6 - 7 | 1449 | 73 | |
NM_001289489.1 | NP_001276418.1 | 5 - 6 | 1344 | 73 | |
NM_028807.4 | NP_083083.3 | 5 - 6 | 1593 | 73 | |
GenBank mRNA | AK004674.1 | - | 6 - 7 | 1450 | 73 |
AK005443.1 | - | 4 - 5 | 1340 | 73 | |
BC042799.1 | - | 2 - 3 | 333 | 73 | |
BC075664.1 | - | 5 - 6 | 1434 | 73 |