The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165980.1 | NP_001159452.1 | 2 - 3 | 485 | 114 |
NM_001165981.1 | NP_001159453.1 | 2 - 3 | 485 | 114 | |
NM_001165982.1 | NP_001159454.1 | 2 - 3 | 485 | 114 | |
NM_198005.2 | NP_932122.2 | 2 - 3 | 485 | 114 | |
GenBank mRNA | AK014717.1 | - | 2 - 3 | 449 | 114 |
AK032671.1 | - | 2 - 3 | 447 | 114 | |
AK079462.1 | - | 2 - 3 | 279 | 114 | |
AK081839.1 | - | 2 - 3 | 484 | 114 | |
AK160679.1 | - | 2 - 3 | 452 | 114 | |
BC031546.1 | - | 2 - 3 | 377 | 114 |