Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tasp1
Entrez Gene ID
75812
Gene Name
taspase, threonine aspartase 1
Gene Aliases
4930485D02Rik, AW986064
Chromosome Location
Chr.2: 139833479 - 140067666 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159640.2 NP_001153112.1 11 - 12 1103 70
NM_175225.5 NP_780434.1 11 - 12 1370 70
NR_110350.1 10 - 11 1277 70
NR_110351.1 11 - 12 1090 70
NR_110352.1 10 - 11 1021 70
GenBank mRNA AK015624.1 - 11 - 12 1090 70
AK036512.1 - 11 - 12 1326 70
AK132709.1 - 10 - 11 1007 70
AK160983.1 - 10 - 11 1238 70
BC024597.1 - 11 - 12 1090 70

View Full Product Details