Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tasp1
Entrez Gene ID
75812
Gene Name
taspase, threonine aspartase 1
Gene Aliases
4930485D02Rik, AW986064
Chromosome Location
Chr.2: 139833479 - 140067666 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159640.2 NP_001153112.1 3 - 4 329 83
NM_001159641.2 NP_001153113.1 3 - 4 329 83
NM_001289611.1 NP_001276540.1 3 - 4 329 83
NM_175225.5 NP_780434.1 3 - 4 596 83
NR_110350.1 3 - 4 596 83
NR_110351.1 3 - 4 329 83
GenBank mRNA AK015624.1 - 3 - 4 329 83
AK036512.1 - 3 - 4 552 83
AK146320.1 - 3 - 4 312 83
AK160983.1 - 3 - 4 557 83
BB665115.1 - 3 - 4 305 83
BC024597.1 - 3 - 4 316 83

View Full Product Details