Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcp11l1
Entrez Gene ID
320554
Gene Name
t-complex 11 like 1
Gene Aliases
C130096D04Rik
Chromosome Location
Chr.2: 104678008 - 104712169 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK047525.1 - 1 - 2 197 103

View Full Product Details