Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sp3
Entrez Gene ID
20687
Gene Name
trans-acting transcription factor 3
Gene Aliases
D130027J01Rik
Chromosome Location
Chr.2: 72936430 - 72980446 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018042.3 NP_001018052.1 1 - 2 200 58
GenBank mRNA BC079874.1 - 1 - 2 200 58

View Full Product Details