Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sp3
Entrez Gene ID
20687
Gene Name
trans-acting transcription factor 3
Gene Aliases
D130027J01Rik
Chromosome Location
Chr.2: 72936430 - 72980446 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018042.3 NP_001018052.1 6 - 7 2221 143
NM_001098425.1 NP_001091895.1 4 - 5 2130 143
GenBank mRNA AF062567.1 - 4 - 5 1857 143
AK004607.1 - 3 - 4 1871 143
AK088238.1 - 4 - 5 2105 143
AK132702.1 - 4 - 5 1876 143
BC027797.1 - 3 - 4 1343 143
BC079874.1 - 6 - 7 2221 143

View Full Product Details