Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agps
Entrez Gene ID
228061
Gene Name
alkylglycerone phosphate synthase
Gene Aliases
5832437L22, 9930035G10Rik, AW123847, Adaps, Adas, Adhaps, Adps, Aldhpsy, bs2
Chromosome Location
Chr.2: 75832177 - 75931350 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172666.3 NP_766254.2 12 - 13 1360 157
GenBank mRNA AK031049.1 - 12 - 13 1362 157
AK031154.1 - 12 - 13 1201 157
AK042745.1 - 12 - 13 1200 157
AK053686.1 - 12 - 13 1349 157
AK079447.1 - 11 - 12 1040 157
BC063086.1 - 12 - 13 1345 157

View Full Product Details