Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr38
Entrez Gene ID
76646
Gene Name
WD repeat domain 38
Gene Aliases
1700123D08Rik
Chromosome Location
Chr.2: 38998309 - 39001588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029687.3 NP_083963.1 5 - 6 590 71
GenBank mRNA AK007246.1 - 5 - 6 581 71
AK077027.1 - - 838 71
BC115629.1 - 5 - 6 568 71

View Full Product Details