Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Accsl
Entrez Gene ID
381411
Gene Name
1-aminocyclopropane-1-carboxylate synthase (non-functional)-like
Gene Aliases
Gm1967
Chromosome Location
Chr.2: 93855360 - 93931236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033452.4 NP_001028624.2 1 - 2 135 96
GenBank mRNA AK135828.1 - 1 - 2 82 96
CF913709.1 - 1 - 2 135 96

View Full Product Details