Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Accsl
Entrez Gene ID
381411
Gene Name
1-aminocyclopropane-1-carboxylate synthase (non-functional)-like
Gene Aliases
Gm1967
Chromosome Location
Chr.2: 93855360 - 93931236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033452.4 NP_001028624.2 8 - 9 1125 64
GenBank mRNA AK135828.1 - 8 - 9 1072 64
AK142866.1 - 6 - 7 616 64
AK163963.1 - 8 - 9 1207 64
BC141227.1 - 6 - 7 504 64
BC145551.1 - 6 - 7 504 64
BC145552.1 - 6 - 7 504 64

View Full Product Details