Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hsd17b12
- Entrez Gene ID
-
56348
- Gene Name
- hydroxysteroid (17-beta) dehydrogenase 12
- Gene Aliases
-
2610510O05Rik, AI172963, KIK-I, Kik1
- Chromosome Location
-
Chr.2: 94032697 - 94157909 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC037620.1 |
- |
2 - 3 |
231 |
72 |
View Full Product Details