Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfyve21
Entrez Gene ID
68520
Gene Name
zinc finger, FYVE domain containing 21
Gene Aliases
1110013H04Rik, AW558780, C85416
Chromosome Location
Chr.12: 111814170 - 111828388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026752.4 NP_081028.3 3 - 4 431 74
GenBank mRNA AK003661.1 - 3 - 4 328 74
AK139356.1 - - 366 74
AK155300.1 - 3 - 4 432 74
AK171099.1 - 3 - 4 372 74
AK197357.1 - 1 - 2 42 74
AK216869.1 - 1 - 2 163 74
BC019521.1 - 3 - 4 361 74
BC132248.1 - 3 - 4 331 74
BC132250.1 - 3 - 4 331 74

View Full Product Details