The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177673.1 | NP_001171144.1 | 2 - 3 | 188 | 75 |
NM_001177674.1 | NP_001171145.1 | 2 - 3 | 370 | 75 | |
GenBank mRNA | AK087410.1 | - | 2 - 3 | 172 | 75 |
BY206757.1 | - | 2 - 3 | 286 | 75 |