Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrbp1
Entrez Gene ID
81910
Gene Name
ribosome binding protein 1
Gene Aliases
1700087N07Rik, 5730465C04Rik, ES/130, mKIAA1398, mRRp0, mRRp1.8, mRRp10, mRRp15a, mRRp15b, mRRp16.8, mRRp2, mRRp41, mRRp47, mRRp5.4, p180
Chromosome Location
Chr.2: 143947395 - 144011296 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024281.2 NP_077243.2 21 - 22 4405 87
GenBank mRNA AK019964.1 - 20 - 21 2338 87
BC022640.1 - 3 - 4 276 87

View Full Product Details