The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009086.2 | NP_033112.2 | 1 - 2 | 267 | 111 |
GenBank mRNA | AK052672.1 | - | 1 - 2 | 237 | 111 |
AK083732.1 | - | 1 - 2 | 249 | 111 | |
AK143501.1 | - | 1 - 2 | 204 | 111 | |
AK144616.1 | - | 1 - 2 | 254 | 111 | |
AK156712.1 | - | 1 - 2 | 258 | 111 | |
BC060656.1 | - | 1 - 2 | 197 | 111 | |
U58280.1 | - | 1 - 2 | 231 | 111 |