Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpsf2
Entrez Gene ID
51786
Gene Name
cleavage and polyadenylation specific factor 2
Gene Aliases
100kDa, 2610024B04Rik, AI662483, Cpsf, MCPSF, mKIAA1367
Chromosome Location
Chr.12: 101975765 - 102006430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016856.3 NP_058552.1 13 - 14 2314 83
GenBank mRNA AF012822.1 - 13 - 14 2300 83
BC007163.1 - 11 - 12 1972 83
BC013628.1 - 13 - 14 2289 83

View Full Product Details