Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnks1bp1
Entrez Gene ID
228140
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
Tab182, mKIAA1741
Chromosome Location
Chr.2: 85048012 - 85073048 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081260.2 NP_001074729.1 6 - 7 4407 64
GenBank mRNA AK129436.1 - 2 - 3 3100 64
BC024499.1 - 1 - 2 305 64
BC025943.1 - 1 - 2 1856 64
BC150947.1 - 6 - 7 4327 64

View Full Product Details