The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010406.2 | NP_034536.1 | 17 - 18 | 2326 | 94 |
GenBank mRNA | M35525.1 | - | 17 - 18 | 2281 | 94 |
M35526.1 | - | 17 - 18 | 2279 | 94 |