The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139197.2 | NP_631936.2 | 2 - 3 | 337 | 75 |
GenBank mRNA | AB084789.1 | - | 1 - 2 | 86 | 75 |
AF292399.1 | - | 1 - 2 | 66 | 75 | |
BC106831.2 | - | 1 - 2 | 66 | 75 |