Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ism1
Entrez Gene ID
319909
Gene Name
isthmin 1 homolog (zebrafish)
Gene Aliases
5430433G21Rik, Ism
Chromosome Location
Chr.2: 139675007 - 139758581 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276489.1 NP_001263418.1 4 - 5 1272 76
GenBank mRNA AK077398.1 - 4 - 5 1273 76

View Full Product Details