Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsun6
Entrez Gene ID
74455
Gene Name
NOL1/NOP2/Sun domain family member 6
Gene Aliases
4933403D21Rik, 4933414E04Rik, NOPD1
Chromosome Location
Chr.2: 14991522 - 15054872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165941.1 NP_001159413.1 3 - 4 654 70
NM_001165942.1 NP_001159414.1 2 - 3 541 70
NM_001165943.1 NP_001159415.1 2 - 3 358 70
NM_028950.4 NP_083226.3 2 - 3 541 70
GenBank mRNA AK016629.1 - 2 - 3 359 70
AK016811.1 - 3 - 4 667 70
AK042750.1 - 2 - 3 541 70
AK077778.1 - 2 - 3 425 70
AK132428.1 - 2 - 3 508 70
BC053751.1 - 2 - 3 509 70

View Full Product Details